Learn More
DuPont™ Tyvek™ 400 Disposable Shirt, USDA, FDA, OSHA
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
- Excellent abrasion resistance due its unique, nonwoven material
- Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Size | 2X-Large |
Color | White |
Neck Style | Collar |
Seam Type | Serged |
Closure Type | Snaps |
Cuff Style | Open |
Style | Extends to hip |