Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 Shoe and Boot Covers
GREENER_CHOICE

Protect your feet from dirt, grease and splashes

Manufacturer:  DuPont™ TY450SWH000200LG

 View more versions of this product

Catalog No. 19-715-362


Add to cart

Alert:

Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

This item is not returnable. View return policy

Description

Description

Protect your feet from dirt, grease and splashes

Abrasion- and tear-resistant, lightweight and comfortable, Tyvek offers a superior combination of barrier, durability and breathability.

Shoe Cover

  • Serged seams; elastic opening
  • Tyvek shaft and sole
  • Height: 8.25 in. (21cm)
Boot Cover
  • Serged seams; elastic opening; elastic ankle
  • Tyvek shaft; Tyvek FC skid-resistant sole
  • Height: 18 in. (46cm)

Specifications

Specifications

Shoe Covers
Polyethylene
Elastic
5.7mil
20.95cm
Standard
White
Serged
8.25 in.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit