Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanWear™ Tyvek™ Round Covers

Durable, form-fitting cover for small bowls, flasks or other consumables.

Manufacturer:  CleanWear™ 10375

 View more versions of this product

Catalog No. 18-892-114


Add to cart

Description

Description

  • Tyvek (style 1 4224) material with melting point 135°C
  • 100% Spun Polyester thread
  • Fulflex textile elastic
Specifications

Specifications

White
Tyvek (Style 1422A)
Round covers
4 in.
Covering small consumables in labs or production areas
40
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit