Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U 89843A, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive allosteric modulator of GABAA receptors. Enhances GABA-induced Cl- currents in the α1β2γ2, α3β2γ2 and α6β2γ2 subtypes. Causes sedation in vivo following i.v. administration without losing "righting reflex". Also displays antioxidant activity.
Specifications
Quantity | 50 mg |
Formula Weight | 321.85 |
Percent Purity | >99% |
Chemical Name or Material | U 89843A |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature