Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U 89843A, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive allosteric modulator of GABAA receptors. Enhances GABA-induced Cl- currents in the α1β2γ2, α3β2γ2 and α6β2γ2 subtypes. Causes sedation in vivo following i.v. administration without losing "righting reflex". Also displays antioxidant activity.
Specifications
Quantity | 10 mg |
Formula Weight | 321.85 |
Percent Purity | >99% |
Chemical Name or Material | U 89843A |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction