Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U 90042, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
GABAA receptor ligand that binds with comparable affinities to three receptor subtypes: α1β2γ2, α3β2γ2 and α6β2γ2 (Ki values are 7.8, 9.5 and 11.0 nM respectively). Potentiates GABA-induced chloride currents in α6β2γ2 receptors. Sedative/hypnotic compound.
Specifications
Quantity | 50 mg |
Formula Weight | 352.78 |
Percent Purity | >99% |
Chemical Name or Material | U 90042 |
Safety and Handling
Recommended Storage : Store at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title