Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U 93631, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
152273-12-6
C17H21N3O2
299.37
MFCD10566712
NXBSEJKZKXIYMD-UHFFFAOYSA-N
tert-butyl 4,4-dimethyl-5h-imidazo 1,5-a quinoxaline-3-carboxylate, 4,5-dihydro-4,4-dimethyl-imidazo 1,5-a quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester, imidazo 1,5-a quinoxaline-3-carboxylic acid, 4,5-dihydro-4,4-dimethyl-, 1,1-dimethylethyl ester, 4-dimethyl-3-t-butylcarboxyl-4,5-dihydro 1,5-a imidazoquinoxaline
197626
tert-butyl 4,4-dimethyl-4H,5H-imidazo[1,5-a]quinoxaline-3-carboxylate
CC(C)(C)OC(=O)C1=C2N(C=N1)C1=CC=CC=C1NC2(C)C
This item is not returnable. View return policy
152273-12-6
C17H21N3O2
299.37
MFCD10566712
NXBSEJKZKXIYMD-UHFFFAOYSA-N
tert-butyl 4,4-dimethyl-5h-imidazo 1,5-a quinoxaline-3-carboxylate, 4,5-dihydro-4,4-dimethyl-imidazo 1,5-a quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester, imidazo 1,5-a quinoxaline-3-carboxylic acid, 4,5-dihydro-4,4-dimethyl-, 1,1-dimethylethyl ester, 4-dimethyl-3-t-butylcarboxyl-4,5-dihydro 1,5-a imidazoquinoxaline
197626
tert-butyl 4,4-dimethyl-4H,5H-imidazo[1,5-a]quinoxaline-3-carboxylate
CC(C)(C)OC(=O)C1=C2N(C=N1)C1=CC=CC=C1NC2(C)C

GABAA receptor antagonist

GABAA receptor antagonist that binds the picrotoxin site and stabilizes the inactive form of the channel via allosteric interaction. Accelerates the decay of GABA-induced Cl- currents with little effect on peak amplitude. Also inhibits 5-HT3A receptors via a similar mechanism.
Quantity 10 mg
Formula Weight 299.37
Percent Purity >99%
Chemical Name or Material U 93631

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit