Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U 93631, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
152273-12-6
C17H21N3O2
299.37
MFCD10566712
NXBSEJKZKXIYMD-UHFFFAOYSA-N
tert-butyl 4,4-dimethyl-5h-imidazo 1,5-a quinoxaline-3-carboxylate, 4,5-dihydro-4,4-dimethyl-imidazo 1,5-a quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester, imidazo 1,5-a quinoxaline-3-carboxylic acid, 4,5-dihydro-4,4-dimethyl-, 1,1-dimethylethyl ester, 4-dimethyl-3-t-butylcarboxyl-4,5-dihydro 1,5-a imidazoquinoxaline
197626
tert-butyl 4,4-dimethyl-4H,5H-imidazo[1,5-a]quinoxaline-3-carboxylate
CC(C)(C)OC(=O)C1=C2N(C=N1)C1=CC=CC=C1NC2(C)C
This item is not returnable. View return policy
152273-12-6
C17H21N3O2
299.37
MFCD10566712
NXBSEJKZKXIYMD-UHFFFAOYSA-N
tert-butyl 4,4-dimethyl-5h-imidazo 1,5-a quinoxaline-3-carboxylate, 4,5-dihydro-4,4-dimethyl-imidazo 1,5-a quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester, imidazo 1,5-a quinoxaline-3-carboxylic acid, 4,5-dihydro-4,4-dimethyl-, 1,1-dimethylethyl ester, 4-dimethyl-3-t-butylcarboxyl-4,5-dihydro 1,5-a imidazoquinoxaline
197626
tert-butyl 4,4-dimethyl-4H,5H-imidazo[1,5-a]quinoxaline-3-carboxylate
CC(C)(C)OC(=O)C1=C2N(C=N1)C1=CC=CC=C1NC2(C)C

GABAA receptor antagonist

GABAA receptor antagonist that binds the picrotoxin site and stabilizes the inactive form of the channel via allosteric interaction. Accelerates the decay of GABA-induced Cl- currents with little effect on peak amplitude. Also inhibits 5-HT3A receptors via a similar mechanism.
Quantity 50 mg
Formula Weight 299.37
Percent Purity >99%
Chemical Name or Material U 93631

Recommended Storage : Store at Room Temperature