Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ u-Plasminogen Activator/Urokinase Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$152.22 - $344.00
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 5328 |
Molecular Weight | TMW: 47.4kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2523605
|
Novus Biologicals™
NBP2523600.005MG |
0.005mg |
Each for $152.22
|
|
NBP2523602
|
Novus Biologicals™
NBP2523600.02MG |
0.02mg |
Each for $344.00
|
|
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 21-431 of Human u-Plasminogen Activator/Urokinase The Recombinant Human u-Plasminogen Activator/Urokinase Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
Phosphate buffered saline (pH 7.4) containing 10% glycerol | |
TMW: 47.4kDa | |
Baculovirus | |
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator | |
Measured by its ability to cleave a peptide substrate Z-GGR-AMC. The specific activity is >7,000 pmol/min/ug. | |
Yes |
SDS-Page | |
5328 | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
u-Plasminogen Activator/Urokinase | |
Unlabeled |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title