Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

u-Plasminogen Activator/Urokinase Mouse anti-Human, Alexa Fluor™ 350, Clone: U-6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223504AF350

Catalog No. NP22304A30


Add to cart

Description

Description

u-Plasminogen Activator/Urokinase Monoclonal antibody specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

u-Plasminogen Activator/Urokinase
U-6
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
PLAU
Native human uPA
Protein A or G purified
Cancer
Primary
5328.0
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone U-6 is specific for human uPA. No reaction with human tissue plasminogen activator (tPA) is seen when tested by ELISA, immunoblotting and enzyme inhibition. No reaction with any other human plasma proteins is seen when tested by immunoblotting.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit