Learn More
u-PlasminogenActivator/Urokinase Mouse anti-Human, Janelia Fluor 646, Clone: U-5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223503JF646
Catalog No. NBP23503646
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
u-PlasminogenActivator/Urokinase Monoclonal antibody specifically detects u-PlasminogenActivator/Urokinase in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
u-PlasminogenActivator/Urokinase
U-5
Immunohistochemistry
Purified
PLAU
Native human uPA
Protein A or G purified
Cancer
Primary
5328
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone U-5 is specific for human uPA. No reaction with human tissue plasminogen activator (tPA) is seen when tested by ELISA, immunoblotting and enzyme inhibition. No reaction with any other human plasma proteins is seen when tested by immunoblotting.