Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Faceshields
Click to view available options
For Use With (Equipment):
Double matrix faceshield headgear (483000)
Matrix faceshield headgear
Matrix headgear
Lens Material:
Acetate
Polycarbonate
Polyester
Description
Available in an assortment of brackets and faceshield sizes and materials.
Compliance
Meets ANSI Z87.1+ standards.
Specifications
Thickness (Metric) | 0.1 cm |
Certifications/Compliance | ANSI Z87.1+, CSA Z94.1 |
Lens Tint | Clear |
For Use With (Equipment) | Matrix headgear |
No. per Case | 48/Cs. |
Lens Material | Polycarbonate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction