Learn More
MCR Safety SafetyToes™ Slipp-R™ Steel-Toe Safety Overshoes
100% rubber steel-toe safety overshoes feature an integrated steel toe, providing safety and durability.
Manufacturer:  MCR Safety 87008
 View more versions of this product
Catalog No. 19-825-560
Add to cart
Description
Description
U.S. Safety SafetyToes Slipp-R Steel-Toe Safety Overshoes resist slips and cuts, keeping you on your toes and safer on the job.
  • Flexible at all temperatures and easy to slip on
  • Won't stiffen or crack
  • Resistant to chemicals, acids and animal fats
  • 100% waterproof
  • Nonmarking
Compliance
Meet OSHA requirements for toe protection; 29 CFR 1910.136(b)(1). Passes ASTM F 2413-05 Paragraph 5.1.4.1(1) Impact Test and Paragraph 5.2.4.1(1) Compression Test in accordance with ASTM F 2412-05 Standard Test Method for Foot Protection.
Specifications
Specifications
Various
Rubber
Includes the most popular size range
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Product should not be used where metatarsal protection is required or where there is potential for electric shocks or objects piercing the sole.