U.S. Safety™ SafetyToes™ Slipp-R™ Steel-Toe Safety Overshoes
100% rubber steel-toe safety overshoes feature an integrated steel toe, providing safety and durability.
Manufacturer: MCR SAFETY COMPANY 87006
 View more versions of this product
Catalog No. 19-825-558
  Check Availability
Add to Cart
Disclaimers:
Product should not be used where metatarsal protection is required or where there is potential for electric shocks or objects piercing the sole.
    Specifications
    Material Rubber
    Size XL (Men's 12-1/2, Women's 14)
    Color Blue
  • Description & Specifications
Related Products
Product Content Feedback