Learn More
CERILLIANT™ U-51754 Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco U0051ML
Catalog No. 01-002-127
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
U-51754 is a highly potent opioid agonist, not structurally related to fentanyl, that has been confirmed in opioid overdoses and deaths. Novel synthetic opioids (NSOs) created to circumvent drug control laws such as U-51754 have no recognized therapeutic use, are clandestinely manufactured and sold on conventional or dark web. This Certified Spiking Solution™ is offered as DEA-exempt solution standard and applicable for use in urine drug testing, clinical toxicology, or forensic analysis applications by LC-MS/MS or GC/MS.
Specifications
Specifications
67-56-1,121279-74-1
9.7°C (49.5°F) - closed cup
C17H24Cl2N2O · HCl
1 mL
1 mg/mL in methanol (as free base)
379.75
Ampule
-20°C
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation