Learn More
CERILLIANT™ U-50488-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco U0081ML
Catalog No. 01-002-130
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This stable-labeled internal standard is suitable for quantitation of U-50488 levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, or isotope dilution methods. U-50488 is a potent opioid agonist, not structurally related to fentanyl. U-50488 belongs to the class of novel synthetic opioids (NSOs) created to circumvent drug control laws with no recognized therapeutic use. NSOs are clandestinely manufactured and sold on conventional or dark web.
Specifications
Specifications
67-56-1
9.7°C (49.5°F) - closed cup
C19H23D3Cl2N2O · HCl
1 mL
100 μg/mL in methanol (as free base)
408.81
Ampule
-20°C
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation