Learn More
U2AF1L4 Mouse anti-Human, Clone: OTI7B12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810433
Catalog No. 50-167-0070
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RNA-binding protein that function as a pre-mRNA splicing factor. Plays a critical role in both constitutive and enhancer-dependent splicing by mediating protein-protein interactions and protein-RNA interactions required for accurate 3 ft.-splice site selection. Acts by enhancing the binding of U2AF2 to weak pyrimidine tracts. Also participates in the regulation of alternative pre-mRNA splicing. Activates exon 5 skipping of PTPRC during T cell activation; an event reversed by GFI1. Binds to RNA at the AG dinucleotide at the 3 ft.-splice site.
Specifications
Specifications
U2AF1L4
Monoclonal
Unconjugated
U2AF1L4
Lyophilized
U2AF1L4
Full length human recombit protein of human U2AF1L4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI7B12
PBS with 8% trehalose and no preservative; pH 7.3
Q8WU68
U2AF1-RS3, U2AF1L3, U2AF1L3V1, U2AF1RS3, U2af26
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
199746