Learn More
U2AF65 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156241AP
Catalog No. 50-173-0360
Add to cart
Description
Description
U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.
Specifications
Specifications
U2AF65
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26368, P26369
hU2AF(65), hU2AF65, U2AF2, U2AF65
Rabbit
IgG
150 μL
-20°C
Primary
11338, 22185, 308335
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
U2AF2
Liquid
U2af2
U2AF65 Fusion Protein Ag8070
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat