Learn More
BioVision IncSupplier Diversity Partner Albumin-to-Creatinine Ratio (ACR): Urine Albumin-to-Creatinine Ratio (UACR) Assay Kit: Only assay kit on the market to measure urine albumin-to-creatinine ratio (UACR). Simple & Convenient. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K551100
Urine Albumin-to-Creatinine Ratio (UACR) Assay Kit: is one of the two markers used to determine chronic kidney disease (CKD). UACR is recommended to be measured on regular basis on people living with Type I and Type II diabetes. UACR is defined as the ratio between albumin (reported in mg/dl) and creatinine (reported in g/dl). This ratio estimates the amount of albumin excreted in urine during a 24 hr period. BioVision's UACR Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects albumin (detection range: 0.02- 2.5 mg/ml), creatinine (detection range: 0.002 -0.5 mg/ml) and UACR. The UACR ratio is determined in two steps: First, albumin is determined by using a probe (AB580) that specifically recognizes albumin (Ex/Em = 600/630 nm). Second, creatinine is converted to sarcosine via enzymatic reactions. Sarcosine is specifically oxidized generating a product that reacts with a probe producing a chromophore that can be detected at 570 nm.
Catalog No. 50-111-9765