Tocris™ UAMC 00039 dihydrochloride

Dipeptidyl peptidase II (DPP-II) inhibitor

Manufacturer: Tocris™ 4954/10

 View more versions of this product

Catalog No. 49-541-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors UAMC 00039 dihydrochloride
    Purity >98
    Molecular Weight 382.76
    Quantity 10mg
    Product Type UAMC 00039 dihydrochloride

    Potent inhibitor of dipeptidyl peptidase II (DPP-II) (IC50 = 0.48nM). Exhibits selectivity for DPP-II against DPP-9, DPP-8 and DPP-IV (IC50 values are 78.6, 142 and 165µM, respectively). Orally available.

    • Chemical Name: (2S)-2-Amino-4-[[(4-chlorophenyl)methyl]amino]-1-(1-piperidinyl)-1-butanone dihydrochloride
    • Formula: C16H24ClN3O.2HCl
    • CAS Number: 697797-51-6
    • Solubility: Soluble to 100mM in water and to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit