Learn More
UBA2 Mouse anti-Canine, Human, Rat, Clone: OTI4A3, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA502504S
Catalog No. 50-167-3557
Add to cart
Description
Description
UBA2 is involved in covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respect to size and ubiquity. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied extensively. Recently, a new group of ubiquitin-like proteins has been receiving a lot of attention. SUMO, or Sentrin, is one of the most intriguing. SUMO has been implicated in the stabilization of target proteins and/or their localization to sub-cellular complexes.The conjugation of SUMO-1, SUMO-2 and SUMO-3 onto target proteins requires the concerted action of the specific E1-activating enzyme SAE1/SAE2, the E2-conjugating enzyme Ubc9, and an E3-like SUMO ligase.
Specifications
Specifications
UBA2
Monoclonal
Unconjugated
UBA2
Liquid
UBA2
Full length human recombit protein of human UBA2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4A3
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UBT2
ARX, HRIHFB2115, SAE2
Mouse
IgG2a
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10054, 308508, 476490