Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBA3 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1B9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA811881

Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E1 ubiquitin-activating enzyme family. The encoded enzyme associates with AppBp1, an amyloid beta precursor protein binding protein, to form a heterodimer, and then the enzyme complex activates NEDD8, a ubiquitin-like protein, which regulates cell division, signaling and embryogenesis. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
UBA3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8C878, Q8TBC4, Q99MI7 | |
UBA3 | |
Human recombit protein fragment corresponding to amino acids 1-463 of human UBA3 produced in E.coli. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
OTI1B9 | |
Unconjugated | |
UBA3 | |
NAE2, UBE1C, hUBA3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
117553, 22200, 476560, 9039 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title