Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBAP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548447

Catalog No. PA548447

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene contains a UBA (ubiquitin associated) domain, which is characteristic of proteins that function in the ubiquitination pathway. This gene may show increased expression in the adrenal gland and lymphatic tissues. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Sep 2013]
Specifications

Specifications

UBAP2
Polyclonal
PBS with 0.09% sodium azide
Q5T6F2, Q91VX2
Ubiquitin-associated protein 2, UBAP-2, UBAP2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55833, 68926
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UBAP2
Liquid
UBAP2
KLH conjugated synthetic peptide between 144-173 amino acids from the N-terminal region of human UBAP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit