Learn More
UBASH3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158231AP
Catalog No. 50-173-0509
Add to cart
Description
Description
This antibody is UBASH3A-specific.
TULA (T cell ubiquitin ligand, UBASH3A, ubiquitin associated and SH3 domain containing A), STS-2 or CLIP4, is a 661 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one SH3 domain and one UBA domain. Expressed at high levels in thymus, bone marrow, spleen and peripheral blood leukocytes, TULA exists as either a homodimer or a homo-oligomer that interferes with the degradation of receptor-type tyrosine kinases and promotes the accumulation of activated receptors on the cell surface. Additionally, TULA is part of an EGFR- and Cbl-containing complex that interacts with ubiquitinated proteins. The gene encoding TULA, which maps to human chromosome 21, may be involved in the pathogenesis of type I diabetes. Multiple isoforms of TULA are produced due to alternative splicing events.
Specifications
Specifications
UBASH3A
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P57075, Q3V3E1
Cbl interacting protein 4, CLIP4, STS 2, STS2, T cell ubiquitin ligand, TULA, UBASH3A
Rabbit
IgG
150 μL
-20°C
Primary
309666, 328795, 53347
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
UBASH3A
Liquid
Ubash3a
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat