Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBB Rabbit anti-Human, Unconjugated, Clone: UBB/3143R, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation RAB00651
Description
UBB Monoclonal antibody specifically detects UBB in Human samples. It is validated for IHC-P.Specifications
UBB | |
Monoclonal | |
Unconjugated | |
UBB | |
Rabbit | |
100 μg | |
7314 | |
Store at -20 to -80°C | |
IgG |
Immunohistochemistry (Paraffin) | |
UBB/3143R | |
ubiquitin B | |
FLJ25987, MGC8385 | |
Recombinant protein corresponding to amino acids 1-119 of human UBB | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction