Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH10/UBE2C Mouse anti-Human, Clone: CPTC-UBE2C-1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen UbcH10/UBE2C
Clone CPTC-UBE2C-1
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122481
View Documents
Novus Biologicals
NBP30750410UG
100 μg
Each for $352.89
Add to cart
 
NB122482
View Documents
Novus Biologicals
NBP30750420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

UbcH10/UBE2C Monoclonal antibody specifically detects UbcH10/UBE2C in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

UbcH10/UBE2C
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
Human
cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C
Recombinant human full-length UbcH10/UBE2C protein
Monoclonal
Protein A or G purified
CPTC-UBE2C-1
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators
10 mM PBS with 0.05% BSA
11065
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit