Learn More
UbcH3/Cdc34 Mouse anti-Human, Alexa Fluor 700, Clone: 90.12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB500110AF700
Catalog No. NBV110AF700
Add to cart
Description
Description
UBE2R1/CDC34 Monoclonal antibody specifically detects UBE2R1/CDC34 in Human samples. It is validated for Western Blot.
Specifications
Specifications
UbcH3/Cdc34
90.12
Western Blot
P49427
cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Purified
CDC34
Full-length human Cdc34 expressed in E. coli.
Protein G purified
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
997