Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH5a/UBE2D1 Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299796B

Catalog No. NB062953


Add to cart

Description

Description

UbcH5a/UBE2D1 Polyclonal antibody specifically detects UbcH5a/UBE2D1 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

UbcH5a/UBE2D1
Biotin
PBS
E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1
Produced in rabbits immunized with purified, recombinant UbcH5a/UBE2D1 (P51668; Ala2-Met147).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Cancer, Hypoxia
Primary
7321
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit