Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH8/Ube2L6 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300032AF350
Description
UbcH8/Ube2L6 Polyclonal specifically detects UbcH8/Ube2L6 in Human samples. It is validated for Western Blot, ELISA.Specifications
UbcH8/Ube2L6 | |
Polyclonal | |
Western Blot, ELISA | |
EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6 | |
Produced in rabbits immunized with purified, recombinant UbcH8/Ube2L6 (O14933-1; Met1-Ser153). | |
0.1 mL | |
Stem Cell Markers | |
9246 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction