Learn More
UBD Rabbit anti-Human, Unconjugated, Clone: HID-21, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20828
Catalog No. 89-337-228
Add to cart
Description
Description
UBD Monoclonal antibody specifically detects UBD in Human samples. It is validated for ICC, IHC, WB-Ce, IF.
Specifications
Specifications
UBD
HID-21
ubiquitin D
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
UBD
FAT10, GABBR1, UBD-3
A synthetic peptide corresponding to human UBD
100 μg
Primary
10537