Learn More
UBE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159121AP
Catalog No. 50-173-0569
Add to cart
Description
Description
UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5 ft. UTR.
Specifications
Specifications
UBE1
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22314, Q02053, Q5U300
A1S9, A1S9T, A1ST, AMCX1, GXP1, Protein A1S9, SMAX2, UBA1, UBA1A, UBE1, UBE1X, Ubiquitin activating enzyme E1
Rabbit
IgG
150 μL
-20°C
Primary
22201, 314432, 7317
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UBA1
Liquid
Uba1
UBE1 Fusion Protein Ag8703
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat