Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2C Mouse anti-Human, Clone:1F5D3, Abnova™
Mouse monoclonal antibody raised against recombinant human UBE2C.
Supplier: Abnova Corporation MAB16614
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for the destruction of mitotic cyclins and for cell cycle progression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]Specifications
UBE2C | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. |
|
UBE2C | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1F5D3 | |
ubiquitin-conjugating enzyme E2C | |
In PBS (0.05% sodium azide) | |
UBCH10, dJ447F3.2 | |
Recombinant protein corresponding to amino acid 1-179 of human UBE2C from E. coli. | |
11065 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction