Learn More
Invitrogen™ UBE2D4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121187
Description
Positive test controls include: BT-474, HepG2, MCF7, Mouse liver, Mouse skeletal muscle, Mouse kidney, Rat lung, Rat liver. Immunogen sequence: MALKRIQKEL TDLQRDPPAQ CSAGPVGDDL FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TKIYHPNINS NGSICLDILR SQWSPALTVS KVLLSICSLL CDPNPDDPLV PEIAHTYKAD REKYNRLARE WTQKYAM.
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro able to promote polyubiquitination using all 7 ubiquitin Lys residues, but may prefer 'Lys-11' and 'Lys-48'-linked polyubiquitination.
Specifications
UBE2D4 | |
Polyclonal | |
Unconjugated | |
Ube2d4 | |
E2 ubiquitin-conjugating enzyme D2B; E2 ubiquitin-conjugating enzyme D4; E2(17)KB 2B; HBUCE1; RGD69425; UBCH5D; Ube2d2; Ube2d2b; UBE2D4; Ubiquitin carrier protein D2B; ubiquitin carrier protein D4; ubiquitin conjugating enzyme E2 D4 (putative); ubiquitin conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme E2 D2B; Ubiquitin-conjugating enzyme E2 D4; ubiquitin-conjugating enzyme E2(17)KB 2B; Ubiquitin-conjugating enzyme E2-17 kDa 2B; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme HBUCE1; ubiquitin-protein ligase D2B; Ubiquitin-protein ligase D4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51619, 546638, 79435 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.66 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70711, Q9Y2X8 | |
Ube2d4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human UBE2D4 (NP_057067.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.