Learn More
UBE2D4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154751AP
Catalog No. 50-173-0258
Add to cart
Description
Description
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro able to promote polyubiquitination using all 7 ubiquitin Lys residues, but may prefer  ft.Lys-11 ft. and  ft.Lys-48 ft.-linked polyubiquitination.
Specifications
Specifications
UBE2D4
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y2X8
HBUCE1, UBCH5D, UBE2D4, Ubiquitin carrier protein D4, Ubiquitin protein ligase D4
Rabbit
IgG
150 μL
-20°C
Primary
51619, 546638, 79435
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UBE2D4
Liquid
UBE2D4
UBE2D4 Fusion Protein Ag7779
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat