Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2H Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548806

Catalog No. PA548806

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and mouse based on sequence homology.

Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes  ft.Lys- 11 ft.- and  ft.Lys-48 ft.-linked polyubiquitination. Capable, in vitro, to ubiquitinate histone H2A.
Specifications

Specifications

UBE2H
Polyclonal
PBS with 0.09% sodium azide
P62256
Ubiquitin-conjugating enzyme E2 H, UbcH2, Ubiquitin carrier protein H, Ubiquitin-conjugating enzyme E2-20K, Ubiquitin-protein ligase H, UBE2H
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7328
Western Blot
Unconjugated
UBE2H
Liquid
UBE2H
KLH conjugated synthetic peptide between 42-70 amino acids from the Central region of human UBE2H.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit