Learn More
UBE2J1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI3D1, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504988
Catalog No. 50-167-5557
Add to cart
Description
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum and may contribute to quality control ER-associated degradation by the ubiquitin-proteasome system.
Specifications
Specifications
UBE2J1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9JJZ4, Q9Y385
CGI-76, HSPC153, HSPC205, HSU93243, NCUBE-1, NCUBE1, UBC6, UBC6E, Ubc6p
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
297961, 474993, 51465, 56228
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3D1
Unconjugated
UBE2J1
Liquid
Ube2j1
Full length human recombit protein of human UBE2J1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat