Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2J2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542351
Description
Peptide sequence: GIQLLNGHAP GAVPNLAGLQ QANRHHGLLG GALANLFVIV GFAAFAYTVK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2J2 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.
Specifications
UBE2J2 | |
Polyclonal | |
Unconjugated | |
UBE2J2 | |
1200007B18Rik; 2400008G19Rik; 5730472G04Rik; AL022923; E2 ubiquitin-conjugating enzyme J2; NCUBE2; NCUBE-2; Non-canonical ubiquitin-conjugating enzyme 2; PRO2121; Ubc6; Ubc6p; Ubc6p homolog; UBE2J2; ubiquitin conjugating enzyme 6; ubiquitin conjugating enzyme E2 J2; ubiquitin conjugating enzyme E2, J2; ubiquitin-conjugating enzyme E2 J2; ubiquitin-conjugating enzyme E2, J2; ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J2 homolog; ubiquitin-conjugating enzyme E2J 2; Unknown (protein for MGC:133997); yeast UBC6 homolog; zgc:162164 | |
Rabbit | |
Protein A | |
RUO | |
118424 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N2K1 | |
UBE2J2 | |
Synthetic peptide directed towards the C-terminal of human UBE2J2 (aa 201-250). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction