Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2M/Ubc12 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | UBE2M/Ubc12 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15503520
|
Novus Biologicals
NBP15503520UL |
20 μL |
Each for $152.22
|
|
NBP155035
|
Novus Biologicals
NBP155035 |
100 μL |
Each for $436.00
|
|
Description
UBE2M/Ubc12 Polyclonal specifically detects UBE2M/Ubc12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UBE2M/Ubc12 | |
Polyclonal | |
Rabbit | |
P61081 | |
9040 | |
Synthetic peptides corresponding to UBE2M(ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast)) The peptide sequence was selected from the N terminal of UBE2M. Peptide sequence IKLFSLKQQKKEEESAGGTKGSSKKASAAQLRIQKDINELNLPKTCDISF. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog | |
UBE2M | |
IgG | |
Affinity Purified | |
21 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title