Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2R1/CDC34 Mouse anti-Human, Clone: CPTC-CDC34-2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30751410UG

 View more versions of this product

Catalog No. NB122501


Add to cart

Description

Description

UBE2R1/CDC34 Monoclonal specifically detects UBE2R1/CDC34 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

UBE2R1/CDC34
CPTC-CDC34-2
Unconjugated
10 mM PBS with 0.05% BSA
cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1
Recombinant human full-length UBE2R1/CDC34 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1 Kappa
100 μg
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
997
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit