Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2R1/CDC34 Mouse anti-Human, Clone: CPTC-CDC34-2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30751420UG
Description
UBE2R1/CDC34 Monoclonal specifically detects UBE2R1/CDC34 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
UBE2R1/CDC34 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1 | |
Recombinant human full-length UBE2R1/CDC34 protein | |
20 μg | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
997 | |
Store at 4°C. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Peptide Array | |
CPTC-CDC34-2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction