Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2S Mouse anti-Human, Mouse, Clone: OTI4H3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA505187

Catalog No. 50-167-5720


Add to cart

Description

Description

This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.
Specifications

Specifications

UBE2S
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16763, Q921J4
E2-EPF, E2EPF, EPF5
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27338, 77891
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI4H3
Unconjugated
UBE2S
Liquid
UBE2S
Full length human recombit protein of human UBE2S produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit