Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2U Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551330

Catalog No. PA551330


Add to cart

Description

Description

The antibody detects endogenous levels of total UBE2U protein.

Catalyzes the covalent attachment of ubiquitin to other proteins.
Specifications

Specifications

UBE2U
Polyclonal
Unconjugated
UBE2U
Liquid
UBE2U
Fusion protein of human UBE2U
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q5VVX9
MGC35130, RP4-636O23.1, UBE2U, Ubiquitin carrier protein U
Rabbit
IgG
100 μL
-20°C
Primary
148581
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit