Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE4A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15503820UL

 View more versions of this product

Catalog No. NBP15503820


Add to cart

Description

Description

UBE4A Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

UBE4A
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
UBE4A
Synthetic peptides corresponding to UBE4A(ubiquitination factor E4A (UFD2 homolog, yeast)) The peptide sequence was selected from the middle region of UBE4A. Peptide sequence QYAPQLAEALIKVFVDIEFTGDPHQFEQKFNYRRPMYPILRYMWGTDTYR.
123 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
B0YJB6
E4MGC133315, KIAA0126ubiquitination factor E4A (homologous to yeast UFD2), ubiquitination factor E4A (UFD2 homolog, yeast), UBOX2, UFD2ubiquitin conjugation factor E4 A
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
9354
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit