Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBFD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550156

Catalog No. PA550156


Add to cart

Description

Description

The antibody detects endogenous levels of total UBFD1 protein.

May play a role as NF-kappa-B regulator. Loftus B.J., Genomics 60:295-308(1999). Ota T., Nat. Genet. 36:40-45(2004).
Specifications

Specifications

UBFD1
Polyclonal
Unconjugated
UBFD1
Liquid
UBFD1
Synthesized peptide derived from internal of human UBFD1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14562, Q78JW9
UBPH
Rabbit
IgG
100 μL
-20°C
Primary
28018, 293454, 56061
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit