Learn More
Ubiquilin 2, Mouse anti-Human,Mouse,Rat, Alexa Fluor 700, Clone: 6H9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP225164AF700
Catalog No. NB005322
Add to cart
Description
Description
Ubiquilin 2 Monoclonal specifically detects Ubiquilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
Specifications
Ubiquilin 2
6H9
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, KnockDown Validated
Purified, Conjugated
UBQLN2
Human Ubiquilin 2 expressed in and purified from E. coli [UniProt# Q9UHD9]
Protein G purified
RUO
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal