Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin B Mouse anti-Human, Clone: UBB/2122, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Ubiquitin B |
---|---|
Clone | UBB/2122 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB121802
|
Novus Biologicals
NBP30716420UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Ubiquitin B Monoclonal specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
Ubiquitin B | |
0.2 mg/ml | |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
UBB/2122 | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
7314 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title