Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin B Rabbit anti-Human, Clone: UBB/3143R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | Ubiquitin B |
---|---|
Clone | UBB/3143R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB121804
|
Novus Biologicals
NBP30716520UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
Ubiquitin B Monoclonal specifically detects Ubiquitin B in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Ubiquitin B | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
UBB/3143R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
7314 | |
IgG | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title