Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin Mouse anti-Human, Clone: Ubi-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1510I25UG
Description
Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human samples. It is validated for Immunohistochemistry, Western Blotting.Specifications
Ubiquitin | |
Monoclonal | |
1.0 mg/mL | |
Purified mouse monoclonal antibody IgG1 in buffer containing PBS with 5 mM sodium azide with 50% glycerol. | |
Purified ubiquitin from bovine red blood cells covalently coupled to KLH using glutaraldehyde. | |
25 μg | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
Ubi-1 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction