Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Ubiquitin, Rabbit Polyclonal Antibody

Ubiquitin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$217.00 - $955.00

Specifications

Antigen Ubiquitin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RB9203P
View Documents
Thermo Scientific™
RB9203P
1mL
Each for $955.00
Add to cart
 
RB9203P0
View Documents
Thermo Scientific™
RB9203P0
100μL
Each for $217.00
Add to cart
 
RB9203P1
View Documents
Thermo Scientific™
RB9203P1
500μL
Each for $538.50
Add to cart
 
RB9203R1
View Documents
Thermo Scientific™
RB9203R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
RB9203R7
View Documents
Thermo Scientific™
RB9203R7
7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Thermo Scientific™ Ubiquitin, Rabbit Polyclonal Antibody.

Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by a 26S complex, multicatalytic cytosolic and nuclear protease termed proteasome. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant full-length human ubiquitin protein

Molecular Weight: 8.5kDa

Positive Control: Human brain from patient with Alzheimer's disease

Cellular Localization: Nuclear, cytoplasmic and membrane This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Ubiquitin
Polyclonal
Rabbit
Primary
IgG
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Recombinant full-length human ubiquitin protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit