Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBL4A Mouse anti-Human, Clone: OTI1D4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525415

Catalog No. PIMA525415


Add to cart

Description

Description

UBL4A is a component of the BAT3 complex, a multiprotein complex involved in the post-translational delivery of tail-anchored (TA) membrane proteins to the endoplasmic reticulum membrane. UBL4A is the TA membrane proteins, also named type II transmembrane proteins, contain a single C-terminal transmembrane region. The complex acts by facilitating TA proteins capture by ASNA1/TRC40: it is recruited to ribosomes synthesizing membrane proteins, interacts with the transmembrane region of newly released TA proteins, and transfers them to ASNA1/TRC40 for targeting.
Specifications

Specifications

UBL4A
Monoclonal
0.52 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P11441
Ubiquitin-like 4A, DX254E, DXS254E, G6PD, GDX, UBL4, GET5, MDY2, TMA24
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8266
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D4
Unconjugated
UBL4A
Liquid
UBL4A
Full length human recombit protein of UBL4A produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit